Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003280.2 | NP_003271.1 | 4 | 266 | ||
GenBank mRNA | AF020769.1 | - | 4 | 244 | ||
AK313743.1 | - | 4 | 266 | |||
BC030244.1 | - | 4 | 253 | 5213381 | ||
CR541865.1 | - | 4 | 240 | |||
CR541885.1 | - | 4 | 240 | |||
X07897.1 | - | 4 | 266 |