Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNS1 SIRNA Assays ›
Gene Symbol:
TNS1
Gene Name:
tensin 1
Gene Aliases:
MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS
Chromosome Location:
Chr.2: 217799789 - 217978853 on Build GRCh38
UniGene ID:
Hs.471381
Species:
Human
siRNA ID
HSS110890
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308022.1
NP_001294951.1
13
1045
NM_001308023.1
NP_001294952.1
13
1045
NM_022648.5
NP_072174.3
14
1254
XM_011511712.1
XP_011510014.1
10
910
XM_011511713.1
XP_011510015.1
10
910
XM_011511715.1
XP_011510017.1
10
910
XM_011511723.2
XP_011510025.1
14
1261
XM_017004801.1
XP_016860290.1
10
910
XM_017004802.1
XP_016860291.1
10
910
XM_017004803.1
XP_016860292.1
10
910
XM_017004804.1
XP_016860293.1
10
910
XM_017004805.1
XP_016860294.1
10
910
XM_017004806.1
XP_016860295.1
10
910
XM_017004807.1
XP_016860296.1
10
910
XM_017004808.1
XP_016860297.1
10
910
XM_017004809.1
XP_016860298.1
14
1251
XM_017004810.1
XP_016860299.1
14
1134
XM_017004811.1
XP_016860300.1
14
1261
XM_017004812.1
XP_016860301.1
13
1211
GenBank mRNA
AK001785.2
-
14
1040
AK300345.1
-
14
1251
AK309785.1
-
16
1354
AK310169.1
-
11
1076
BC051304.1
-
11
1121
BC071905.1
-
11
1121
BC116187.1
-
12
1018
40074822
BC116188.1
-
12
1018
40074825
BC126910.1
-
12
1018
BC140942.1
-
14
1186
9056754
BE142921.1
-
6
510
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cell-substrate junction assembly
fibroblast migration
actin binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline