Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001067.3 | NP_001058.2 | 19, 20 | 2450 | ||
XM_005257632.1 | XP_005257689.1 | 19, 20 | 2392 | |||
XM_011525165.1 | XP_011523467.1 | 19, 20 | 2396 | |||
GenBank mRNA | BC013429.1 | - | 19, 20 | 2417 | ||
BC140791.1 | - | 19, 20 | 2320 | 9021659 | ||
J04088.1 | - | 19, 20 | 2327 |