Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001067.3 | NP_001058.2 | 20, 21 | 2596 | ||
XM_005257632.1 | XP_005257689.1 | 20, 21 | 2538 | |||
XM_011525165.1 | XP_011523467.1 | 20, 21 | 2542 | |||
GenBank mRNA | BC013429.1 | - | 20, 21 | 2563 | ||
BC140791.1 | - | 20, 21 | 2466 | 9021659 | ||
J04088.1 | - | 20, 21 | 2473 |