Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
TPM1
Gene Name:
tropomyosin 1 (alpha)
Gene Aliases:
C15orf13, CMD1Y, CMH3, HEL-S-265, HTM-alpha, LVNC9, TMSA
Chromosome Location:
Chr.15: 63042196 - 63071915 on Build GRCh38
UniGene ID:
Hs.133892
Species:
Human
siRNA ID
HSS110927
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000366.5
NP_000357.3
5
697
NM_001018004.1
NP_001018004.1
5
697
NM_001018005.1
NP_001018005.1
5
697
NM_001018006.1
NP_001018006.1
5
697
NM_001018007.1
NP_001018007.1
5
697
NM_001018008.1
NP_001018008.1
4
537
NM_001018020.1
NP_001018020.1
5
697
NM_001301244.1
NP_001288173.1
5
697
NM_001301289.1
NP_001288218.1
4
537
XM_005254637.1
XP_005254694.1
6
778
XM_005254638.3
XP_005254695.2
5
1140
XM_005254639.3
XP_005254696.2
5
1140
XM_005254645.2
XP_005254702.2
5
1140
XM_005254646.2
XP_005254703.1
4
523
XM_005254647.3
XP_005254704.1
4
523
XM_005254650.3
XP_005254707.1
4
523
XM_005254651.1
XP_005254708.1
4
523
XM_005254652.1
XP_005254709.1
4
523
XM_005254653.2
XP_005254710.1
4
523
XM_006720667.3
XP_006720730.2
5
1140
XM_017022534.1
XP_016878023.1
5
1140
XM_017022535.1
XP_016878024.1
5
652
XM_017022536.1
XP_016878025.1
5
1140
XM_017022537.1
XP_016878026.1
5
652
XM_017022538.1
XP_016878027.1
5
1140
XM_017022539.1
XP_016878028.1
5
1140
XM_017022540.1
XP_016878029.1
4
523
XM_017022541.1
XP_016878030.1
4
523
GenBank mRNA
AB209041.1
-
4
574
AF474156.1
-
4
299
AK057989.1
-
7
483
AK092051.1
-
4
1414
AK092573.1
-
3
1519
AK131384.1
-
6
711
AK298625.1
-
4
413
AK299387.1
-
5
586
AK301319.1
-
4
433
AK309170.1
-
5
906
AL050179.1
-
5
578
AY640414.1
-
5
506
AY640415.1
-
5
506
BC007433.2
-
5
647
2962925
BC050473.1
-
4
524
BC053545.1
-
4
535
5730451
BX458841.2
-
5
566
CD679271.1
-
1
28
DQ424903.1
-
4
398
GQ472198.1
-
5
647
KU178366.1
-
5
506
KU178367.1
-
5
506
M19713.1
-
5
562
M19714.1
-
5
448
M19715.1
-
4
343
X12369.1
-
4
336
Z24727.1
-
721
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
movement of cell or subcellular component
muscle contraction
regulation of muscle contraction
cytoskeleton organization
regulation of heart contraction
regulation of cell shape
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
negative regulation of vascular smooth muscle cell proliferation
negative regulation of vascular associated smooth muscle cell migration
actin binding
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
structural constituent of muscle
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0