Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290224.1 | NP_001277153.1 | 13 | 1189 | ||
NM_199259.3 | NP_954868.1 | 17 | 1411 | |||
NM_199260.3 | NP_954869.1 | 16 | 1351 | |||
NM_199261.3 | NP_954870.2 | 18 | 1465 | |||
GenBank mRNA | AF007118.1 | - | 18 | 1413 | ||
AF495908.1 | - | 17 | 1359 | |||
AK308254.1 | - | 17 | 1308 | |||
AL833345.1 | - | 18 | 1372 | |||
AY219886.1 | - | 17 | 1359 | |||
AY219887.1 | - | 16 | 1299 | |||
AY219888.1 | - | 13 | 1137 | |||
BC028719.1 | - | 17 | 1394 | 5269598 |