Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290224.1 | NP_001277153.1 | 17 | 1532 | ||
NM_199259.3 | NP_954868.1 | 21 | 1754 | |||
NM_199260.3 | NP_954869.1 | 20 | 1694 | |||
NM_199261.3 | NP_954870.2 | 22 | 1808 | |||
GenBank mRNA | AF495908.1 | - | 21 | 1702 | ||
AI149736.1 | - | 3 | 409 | |||
AK314285.1 | - | 22 | 1707 | |||
AL833345.1 | - | 22 | 1715 |