Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142407.2 | NP_001135879.1 | 7 | 645 | ||
NM_001142408.2 | NP_001135880.1 | 8 | 702 | |||
NM_001142417.2 | NP_001135889.1 | 8 | 682 | |||
NM_001142435.2 | NP_001135907.1 | 7 | 665 | |||
NM_001261822.1 | NP_001248751.1 | 7 | 671 | |||
NM_003296.3 | NP_003287.1 | 8 | 708 | |||
XM_005249349.1 | XP_005249406.1 | 8 | 682 | |||
XM_005249350.1 | XP_005249407.1 | 7 | 640 | |||
XM_005249351.3 | XP_005249408.1 | 7 | 660 | |||
XM_005249352.3 | XP_005249409.1 | 7 | 651 | |||
XM_005249353.3 | XP_005249410.1 | 8 | 687 | |||
XM_005249356.1 | XP_005249413.1 | 8 | 685 | |||
XM_011514841.1 | XP_011513143.1 | 8 | 1017 | |||
XM_011514842.1 | XP_011513144.1 | 7 | 980 | |||
XM_011514843.2 | XP_011513145.1 | 8 | 687 | |||
GenBank mRNA | AK223337.1 | - | 8 | 645 | ||
AK292385.1 | - | 8 | 686 | |||
AY289796.1 | - | 8 | 652 | |||
AY292862.1 | - | 7 | 642 | |||
AY292863.1 | - | 7 | 646 | |||
AY773479.1 | - | 8 | 688 | |||
BC022011.2 | - | 8 | 655 | 4826427 | ||
BC107707.1 | - | 7 | 602 | 6618679 | ||
BG718726.1 | - | 7 | 641 | |||
BI562149.1 | - | 7 | 627 | |||
BI831679.1 | - | 2 | 57 | |||
BM552843.1 | - | 7 | 580 | |||
BP369727.1 | - | 2 | 159 | |||
M25532.1 | - | 8 | 631 | |||
X95239.1 | - | 8 | 682 |