Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP90B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003299.2 | NP_003290.1 | 12 | 1691 | ||
GenBank mRNA | AB209534.1 | - | 12 | 1587 | ||
AF087988.1 | - | 5 | 495 | |||
AJ890084.1 | - | 12 | 1643 | |||
AK025459.1 | - | 12 | 1643 | |||
AK295262.1 | - | 6 | 613 | |||
AK300521.1 | - | 8 | 864 | |||
AY040226.1 | - | 11 | 1446 | |||
BC066656.1 | - | 12 | 1605 | 6165138 | ||
EU794608.1 | - | 12 | 1605 | |||
FJ460515.1 | - | 12 | 1605 | |||
X15187.1 | - | 12 | 1614 |