Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP90B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003299.2 | NP_003290.1 | 16 | 2410 | ||
GenBank mRNA | AB209534.1 | - | 15 | 2583 | ||
AJ890084.1 | - | 16 | 2362 | |||
AK025459.1 | - | 16 | 2362 | |||
AK295262.1 | - | 10 | 1332 | |||
AK300521.1 | - | 12 | 1583 | |||
AY040226.1 | - | 15 | 2165 | |||
BC066656.1 | - | 16 | 2324 | 6165138 | ||
CA419931.1 | - | 3 | 465 | |||
EU794608.1 | - | 16 | 2324 | |||
FJ460515.1 | - | 16 | 2324 | |||
X15187.1 | - | 16 | 2333 |