Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199427.1 | NP_001186356.1 | 8 | 1577 | ||
NM_003300.3 | NP_003291.2 | 11 | 1826 | |||
NM_145725.2 | NP_663777.1 | 12 | 1965 | |||
NM_145726.2 | NP_663778.1 | 11 | 1890 | |||
XM_011537116.2 | XP_011535418.1 | 10 | 1785 | |||
XM_011537117.2 | XP_011535419.1 | 10 | 1773 | |||
XM_011537118.2 | XP_011535420.1 | 9 | 1693 | |||
XM_017021617.1 | XP_016877106.1 | 12 | 2385 | |||
XM_017021618.1 | XP_016877107.1 | 12 | 3881 | |||
XM_017021619.1 | XP_016877108.1 | 11 | 3808 | |||
GenBank mRNA | AK303172.1 | - | 8 | 1454 | ||
BC075086.2 | - | 10 | 1637 | 30915387 | ||
BC075087.2 | - | 10 | 1637 | 30915599 | ||
BX247977.1 | - | 12 | 1811 | |||
L38509.1 | - | 10 | 1673 | |||
U15637.1 | - | 12 | 1819 | |||
U19260.1 | - | 11 | 1762 | |||
U21092.1 | - | 11 | 1829 |