Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270483.1 | NP_001257412.1 | 2 | 211 | ||
NM_003312.5 | NP_003303.2 | 2 | 243 | |||
GenBank mRNA | AI869664.1 | - | 2 | 322 | ||
AK000862.1 | - | 2 | 237 | |||
AK091874.1 | - | 1 | 863 | |||
AK223521.1 | - | 2 | 237 | |||
AK301300.1 | - | 2 | 237 | |||
BC010148.2 | - | 2 | 220 | 4329727 | ||
BI820468.1 | - | 2 | 240 | |||
BQ231617.1 | - | 2 | 236 | |||
CR456598.1 | - | 1 | 246 | |||
CR456998.1 | - | 1 | 168 | |||
D87292.1 | - | 2 | 216 |