Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282500.1 | NP_001269429.1 | 2 | 253 | ||
NM_003314.2 | NP_003305.1 | 2 | 256 | |||
GenBank mRNA | AK222641.1 | - | 2 | 207 | ||
AK222984.1 | - | 2 | 201 | |||
AK315262.1 | - | 2 | 211 | |||
BC000942.3 | - | 1 | 161 | 3446109 | ||
BP245572.1 | - | 2 | 256 | |||
DN989835.1 | - | 2 | 209 | |||
U46570.1 | - | 2 | 183 |