Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144766.2 | NP_001138238.1 | 12 | 1396 | ||
NM_003315.3 | NP_003306.3 | 12 | 1628 | |||
NR_029431.1 | 11 | 1539 | ||||
XM_011525167.2 | XP_011523469.1 | 13 | 1558 | |||
XM_011525168.2 | XP_011523470.1 | 13 | 1518 | |||
XM_011525169.2 | XP_011523471.1 | 12 | 1370 | |||
XM_017024994.1 | XP_016880483.1 | 13 | 1552 | |||
GenBank mRNA | AK291047.1 | - | 12 | 1329 | ||
AK296602.1 | - | 12 | 1389 | |||
AK298860.1 | - | 12 | 1344 | |||
AK316081.1 | - | 11 | 1265 | |||
BC003601.1 | - | 11 | 1251 | 3543074 | ||
BC011837.2 | - | 12 | 1347 | 3945721 | ||
BC033772.1 | - | 12 | 1373 | 3924081 | ||
BQ445927.1 | - | 3 | 466 | |||
BX647209.1 | - | 12 | 1584 | |||
U46571.1 | - | 12 | 1343 |