Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205296.1 | NP_001192225.1 | 6 | 741 | ||
NM_007344.3 | NP_031370.2 | 7 | 2115 | |||
NR_134525.1 | 7 | 2115 | ||||
XM_006717273.3 | XP_006717336.2 | 7 | 2123 | |||
XM_017015105.1 | XP_016870594.1 | 7 | 2622 | |||
GenBank mRNA | AK302902.1 | - | 7 | 2101 | ||
BC127669.1 | - | 6 | 716 | |||
BC127670.1 | - | 6 | 716 | |||
BC143048.1 | - | 6 | 716 | |||
BC143049.1 | - | 6 | 716 | |||
BC143821.1 | - | 7 | 2088 |