Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TULP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003323.2 | NP_003314.2 | 5 | 392 | ||
XM_006723347.3 | XP_006723410.1 | 5 | 392 | |||
XM_006723348.3 | XP_006723411.1 | 5 | 336 | |||
XM_011527252.2 | XP_011525554.1 | 6 | 496 | |||
XM_011527253.2 | XP_011525555.1 | 5 | 383 | |||
XM_011527254.2 | XP_011525556.1 | 5 | 392 | |||
XM_011527255.2 | XP_011525557.1 | 4 | 340 | |||
XM_011527256.2 | XP_011525558.1 | 3 | 307 | |||
GenBank mRNA | BC026070.2 | - | 5 | 386 | 4837710 | |
U82469.1 | - | 5 | 334 |