Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYRO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006293.3 | NP_006284.2 | 14 | 1974 | ||
XM_017022542.1 | XP_016878031.1 | 14 | 1939 | |||
XM_017022543.1 | XP_016878032.1 | 14 | 1775 | |||
GenBank mRNA | AB209431.1 | - | 14 | 2003 | ||
AK295496.1 | - | 3 | 269 | |||
BC029925.2 | - | 6 | 624 | |||
BC034960.1 | - | 14 | 1756 | |||
BC049368.1 | - | 14 | 1748 | 4821587 | ||
BC051756.1 | - | 14 | 1753 | 6095596 | ||
D17517.1 | - | 14 | 1962 | |||
D50479.1 | - | 14 | 1734 | |||
U02566.1 | - | 11 | 1185 | |||
U05682.1 | - | 14 | 1744 | |||
X72886.1 | - | 3 | 184 |