Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2AF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025203.1 | NP_001020374.1 | 2 | 186 | ||
NM_001025204.1 | NP_001020375.1 | 2 | 186 | |||
NM_006758.2 | NP_006749.1 | 2 | 186 | |||
XM_011529743.2 | XP_011528045.1 | 2 | 899 | |||
XM_017028468.1 | XP_016883957.1 | 2 | 899 | |||
GenBank mRNA | AB451244.1 | - | 2 | 102 | ||
AF370386.1 | - | 1 | 909 | |||
AJ627978.1 | - | 2 | 132 | |||
AK307560.1 | - | 2 | 135 | |||
AL832665.1 | - | 2 | 132 | |||
BC001177.1 | - | 2 | 130 | 3534272 | ||
BC001923.1 | - | 2 | 130 | 2823296 | ||
BC005915.1 | - | 2 | 132 | 4295037 | ||
CB110007.1 | - | 2 | 186 | |||
M96982.1 | - | 2 | 140 |
Gene Symbol: U2AF1L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320646.1 | NP_001307575.1 | 2 | 214 | ||
NM_001320648.1 | NP_001307577.1 | 2 | 214 | |||
NM_001320650.1 | NP_001307579.1 | 2 | 214 | |||
NM_001320651.1 | NP_001307580.1 | 2 | 214 | |||
XM_017028219.1 | XP_016883708.1 | 2 | 214 | |||
XM_017028220.1 | XP_016883709.1 | 2 | 214 | |||
GenBank mRNA | AL522322.3 | - | 2 | 131 | ||
BG612658.1 | - | 2 | 179 | |||
CN280267.1 | - | 2 | 91 | |||
CN280269.1 | - | 2 | 151 | |||
DA211043.1 | - | 2 | 214 |