Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2AF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025203.1 | NP_001020374.1 | 5 | 380 | ||
NM_001025204.1 | NP_001020375.1 | 6 | 447 | |||
NM_006758.2 | NP_006749.1 | 5 | 380 | |||
GenBank mRNA | AB451244.1 | - | 5 | 296 | ||
AF370386.1 | - | 4 | 1103 | |||
AJ627978.1 | - | 5 | 326 | |||
AK098266.1 | - | 1 | 903 | |||
AK307560.1 | - | 5 | 329 | |||
BC001177.1 | - | 5 | 324 | 3534272 | ||
BC001923.1 | - | 5 | 324 | 2823296 | ||
BC005915.1 | - | 6 | 393 | 4295037 | ||
CB110007.1 | - | 5 | 380 | |||
M96982.1 | - | 5 | 334 |
Gene Symbol: U2AF1L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320646.1 | NP_001307575.1 | 5 | 408 | ||
NM_001320648.1 | NP_001307577.1 | 5 | 408 | |||
NM_001320650.1 | NP_001307579.1 | 5 | 379 | |||
NM_001320651.1 | NP_001307580.1 | 6 | 475 | |||
GenBank mRNA | AL522322.3 | - | 5 | 325 | ||
BG612658.1 | - | 5 | 344 | |||
CN280267.1 | - | 6 | 352 | |||
CN280269.1 | - | 5 | 345 |