Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033930.2 | NP_001029102.1 | 4 | 345 | ||
NM_001321017.1 | NP_001307946.1 | 4 | 344 | |||
NM_001321018.1 | NP_001307947.1 | 4 | 971 | |||
NM_001321019.1 | NP_001307948.1 | 4 | 509 | |||
NM_001321020.1 | NP_001307949.1 | 4 | 543 | |||
NM_001321021.1 | NP_001307950.1 | 4 | 358 | |||
NM_001321022.1 | NP_001307951.1 | 3 | 258 | |||
NM_003333.4 | NP_003324.1 | 4 | 323 | |||
XM_005260052.2 | XP_005260109.1 | 4 | 289 | |||
XM_005260053.3 | XP_005260110.1 | 4 | 321 | |||
XM_005260054.2 | XP_005260111.1 | 3 | 251 | |||
XM_006722871.2 | XP_006722934.1 | 4 | 975 | |||
XM_017027198.1 | XP_016882687.1 | 4 | 1298 | |||
GenBank mRNA | AF348700.1 | - | 4 | 270 | ||
AK314717.1 | - | 4 | 255 | |||
AV707734.1 | - | 323 | ||||
BC101830.1 | - | 4 | 287 | 8069336 | ||
BC101832.1 | - | 4 | 287 | 8069338 | ||
BC143668.1 | - | 4 | 287 | 9052182 | ||
BE514609.1 | - | 4 | 490 | |||
BG341101.1 | - | 4 | 361 | |||
BQ428515.1 | - | 3 | 252 | |||
BQ428688.1 | - | 325 | ||||
CB124676.1 | - | 4 | 289 | |||
CD242660.1 | - | 4 | 550 | |||
CD385706.1 | - | 4 | 514 | |||
DA949569.1 | - | 4 | 344 | |||
HY153317.1 | - | 4 | 322 | |||
X56998.1 | - | 3 | 238 | |||
X56999.1 | - | 3 | 247 | |||
Y00361.1 | - | 2 | 113 |