Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202498.1 | NP_001189427.1 | 2 | 95 | ||
NM_003344.3 | NP_003335.1 | 2 | 491 | |||
NM_182697.2 | NP_874356.1 | 2 | 491 | |||
GenBank mRNA | AB451490.1 | - | 2 | 86 | ||
AK316590.1 | - | 2 | 74 | |||
AY302138.1 | - | 2 | 86 | |||
BC006277.2 | - | 2 | 403 | 3957107 | ||
BI088146.1 | - | 2 | 95 | |||
BT006756.1 | - | 2 | 86 | |||
CB995681.1 | - | 508 | ||||
KJ534959.1 | - | 2 | 86 | |||
Z29328.1 | - | 2 | 139 | |||
Z29330.1 | - | 2 | 139 |