Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006952.3 | NP_008883.2 | 6 | 626 | ||
GenBank mRNA | AB002155.1 | - | 6 | 584 | ||
AB015234.1 | - | 5 | 537 | |||
AF042331.1 | - | 5 | 547 | |||
AF082888.1 | - | 5 | 507 | |||
AK225868.1 | - | 5 | 517 | |||
AK303924.1 | - | 5 | 590 | |||
AK313787.1 | - | 6 | 648 | |||
BC063568.1 | - | 5 | 574 | 3861427 | ||
CR457084.1 | - | 5 | 537 |