Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006952.3 | NP_008883.2 | 6 | 693 | ||
GenBank mRNA | AB002155.1 | - | 6 | 651 | ||
AB015234.1 | - | 5 | 604 | |||
AF042331.1 | - | 5 | 614 | |||
AF082888.1 | - | 5 | 574 | |||
AK225868.1 | - | 5 | 584 | |||
AK303924.1 | - | 5 | 657 | |||
AK313787.1 | - | 6 | 715 | |||
BC063568.1 | - | 5 | 641 | 3861427 | ||
CR457084.1 | - | 5 | 604 |