Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012474.4 | NP_036606.2 | 7 | 1059 | ||
XM_011509970.2 | XP_011508272.2 | 5 | 594 | |||
XM_017002236.1 | XP_016857725.1 | 8 | 1264 | |||
GenBank mRNA | AB062451.1 | - | 7 | 678 | ||
AF236637.1 | - | 7 | 738 | |||
AK294537.1 | - | 6 | 589 | |||
BC002906.2 | - | 7 | 899 | 3940564 | ||
BT006860.1 | - | 4 | 270 | |||
CR456857.1 | - | 4 | 270 | |||
D78335.1 | - | 5 | 488 |