Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL14A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021110.3 | NP_066933.1 | 21 | 2844 | ||
XM_005251059.3 | XP_005251116.1 | 21 | 2844 | |||
XM_006716651.3 | XP_006716714.1 | 21 | 2707 | |||
XM_017013809.1 | XP_016869298.1 | 21 | 2633 | |||
GenBank mRNA | AK293080.1 | - | 21 | 2814 | ||
AK307716.1 | - | 21 | 2808 | |||
AK311498.1 | - | 21 | 2808 | |||
BC140893.1 | - | 20 | 2613 | 9021761 | ||
M64108.1 | - | 16 | 2013 |