Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVRAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003369.3 | NP_003360.2 | 12 | 1350 | ||
XM_011545226.2 | XP_011543528.1 | 12 | 1665 | |||
XM_011545227.2 | XP_011543529.1 | 12 | 1665 | |||
XM_011545228.2 | XP_011543530.1 | 12 | 1665 | |||
XM_017018224.1 | XP_016873713.1 | 10 | 1104 | |||
XM_017018225.1 | XP_016873714.1 | 10 | 1104 | |||
XM_017018226.1 | XP_016873715.1 | 12 | 1665 | |||
XM_017018227.1 | XP_016873716.1 | 7 | 691 | |||
GenBank mRNA | AB012958.1 | - | 12 | 1287 | ||
AK095352.1 | - | 6 | 927 | |||
AK222772.1 | - | 12 | 1241 | |||
AK291011.1 | - | 12 | 1328 | |||
AK293782.1 | - | 12 | 1091 | |||
AK296871.1 | - | 6 | 470 | |||
AK311341.1 | - | 8 | 744 | |||
AK316133.1 | - | 6 | 537 | |||
BC064837.1 | - | 12 | 1260 | 6186851 |