Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003373.3 | NP_003364.1 | 10 | 1411 | ||
NM_014000.2 | NP_054706.1 | 10 | 1411 | |||
GenBank mRNA | AK126978.1 | - | 10 | 1666 | ||
AK304823.1 | - | 9 | 1340 | |||
AL832130.1 | - | 3 | 333 | |||
BC039174.1 | - | 10 | 1398 | 4520338 | ||
BX537994.1 | - | 12 | 4121 | |||
EU794673.1 | - | 10 | 1411 | |||
M33308.1 | - | 10 | 1367 |