Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003373.3 | NP_003364.1 | 18 | 2776 | ||
NM_014000.2 | NP_054706.1 | 18 | 2776 | |||
GenBank mRNA | AK126978.1 | - | 18 | 3031 | ||
AK296509.1 | - | 7 | 1047 | |||
AK300280.1 | - | 4 | 557 | |||
AK304823.1 | - | 17 | 2705 | |||
AL832130.1 | - | 11 | 1698 | |||
BC013847.1 | - | 3, 4 | 288 | |||
BC039174.1 | - | 18 | 2763 | 4520338 | ||
BX537994.1 | - | 20 | 5486 | |||
EU794673.1 | - | 18 | 2776 | |||
M33308.1 | - | 18 | 2732 |