Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003380.3 | NP_003371.2 | 3, 4 | 1041 | ||
XM_006717500.1 | XP_006717563.1 | 2, 3 | 2116 | |||
GenBank mRNA | AF328728.1 | - | 2, 3 | 757 | ||
AK056766.1 | - | 4, 5 | 710 | |||
AK091813.1 | - | 3, 4 | 733 | |||
AK093924.1 | - | 1, 2 | 712 | |||
AK097336.1 | - | 3, 4 | 737 | |||
AK098444.1 | - | 3, 4 | 611 | |||
AK222482.1 | - | 2, 3 | 771 | |||
AK222507.1 | - | 2, 3 | 773 | |||
AK222602.1 | - | 2, 3 | 773 | |||
AK290643.1 | - | 2, 3 | 653 | |||
BC000163.2 | - | 2, 3 | 752 | 2985712 | ||
BC030573.2 | - | 2, 3 | 752 | 3637336 | ||
BC066956.1 | - | 3, 4 | 829 | 4823475 | ||
CR407690.1 | - | 2, 3 | 628 | |||
EU794672.1 | - | 2, 3 | 773 | |||
KU178388.1 | - | 2, 3 | 628 | |||
KU178389.1 | - | 3, 4 | 245 | |||
M25246.1 | - | 2, 3 | 132 | |||
X56134.1 | - | 2, 3 | 671 | |||
Z19554.2 | - | 2, 3 | 750 |