Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003380.3 | NP_003371.2 | 4 | 1068 | ||
XM_006717500.1 | XP_006717563.1 | 3 | 2143 | |||
GenBank mRNA | AF328728.1 | - | 3 | 784 | ||
AK056766.1 | - | 5 | 737 | |||
AK091813.1 | - | 4 | 760 | |||
AK093924.1 | - | 2 | 739 | |||
AK097336.1 | - | 4 | 764 | |||
AK098444.1 | - | 4 | 638 | |||
AK222482.1 | - | 3 | 798 | |||
AK222507.1 | - | 3 | 800 | |||
AK222602.1 | - | 3 | 800 | |||
AK290643.1 | - | 3 | 680 | |||
BC000163.2 | - | 3 | 779 | 2985712 | ||
BC030573.2 | - | 3 | 779 | 3637336 | ||
BC066956.1 | - | 4 | 856 | 4823475 | ||
CR407690.1 | - | 3 | 655 | |||
EU794672.1 | - | 3 | 800 | |||
KU178388.1 | - | 3 | 655 | |||
KU178389.1 | - | 4 | 272 | |||
M25246.1 | - | 3 | 159 | |||
X56134.1 | - | 3 | 698 | |||
Z19554.2 | - | 3 | 777 |