Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251882.1 | NP_001238811.1 | 10, 11 | 1409 | ||
NM_001251883.1 | NP_001238812.1 | 6, 7 | 669 | |||
NM_001251884.1 | NP_001238813.1 | 8, 9 | 923 | |||
NM_001251885.1 | NP_001238814.1 | 9, 10 | 986 | |||
NM_004624.3 | NP_004615.2 | 9, 10 | 1032 | |||
XM_005265437.2 | XP_005265494.1 | 9, 10 | 998 | |||
XM_005265438.3 | XP_005265495.1 | 9, 10 | 1561 | |||
XM_005265439.2 | XP_005265496.1 | 8, 9 | 895 | |||
XM_011534079.1 | XP_011532381.1 | 10, 11 | 1760 | |||
XM_011534080.2 | XP_011532382.1 | 9, 10 | 1015 | |||
GenBank mRNA | AK056819.1 | - | 10, 11 | 1409 | ||
AK293548.1 | - | 6, 7 | 664 | |||
AK294609.1 | - | 8, 9 | 923 | |||
AK298115.1 | - | 9, 10 | 955 | |||
AK314334.1 | - | 9, 10 | 1001 | |||
BC064424.1 | - | 9, 10 | 988 | 5752796 | ||
EF577396.1 | - | 9, 10 | 908 | |||
L13288.1 | - | 9, 10 | 1002 | |||
L20295.1 | - | 8, 9 | 812 | |||
X75299.1 | - | 9, 10 | 928 |
Set Membership: |
G-protein coupled receptor |