Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139443.1 | NP_001132915.1 | 8 | 1352 | ||
NM_001300786.1 | NP_001287715.1 | 7 | 1271 | |||
NM_001300787.1 | NP_001287716.1 | 8 | 1352 | |||
NM_004183.3 | NP_004174.1 | 9 | 1540 | |||
NR_134580.1 | 9 | 1743 | ||||
XM_005274210.3 | XP_005274267.1 | 9 | 1650 | |||
XM_005274215.3 | XP_005274272.1 | 7 | 1305 | |||
XM_005274216.3 | XP_005274273.1 | 8 | 1665 | |||
XM_011545229.2 | XP_011543531.1 | 9 | 1090 | |||
XM_011545230.2 | XP_011543532.1 | 8 | 893 | |||
XM_011545233.2 | XP_011543535.1 | 3 | 787 | |||
XM_017018228.1 | XP_016873717.1 | 7 | 1305 | |||
XM_017018229.1 | XP_016873718.1 | 9 | 1853 | |||
XM_017018230.1 | XP_016873719.1 | 7 | 1508 | |||
GenBank mRNA | AB209451.1 | - | 6 | 2581 | ||
AF057169.1 | - | 9 | 1064 | |||
AF057170.1 | - | 9 | 1267 | |||
AF073501.1 | - | 9 | 1048 | |||
AK289681.1 | - | 9 | 1271 | |||
AK293701.1 | - | 8 | 1006 | |||
AK295450.1 | - | 8 | 1086 | |||
AK295998.1 | - | 8 | 886 | |||
AK301392.1 | - | 7 | 869 | |||
AY515704.1 | - | 8 | 960 | |||
BC015220.2 | - | 7 | 1271 | 3877806 | ||
BC041664.1 | - | 8 | 866 | 5194649 | ||
JQ954696.1 | - | 8 | 1026 | |||
KU178390.1 | - | 6 | 699 | |||
KU178391.1 | - | 7 | 780 | |||
KU178392.1 | - | 6 | 627 |