Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000638.3 | NP_000629.3 | 6 | 1099 | ||
GenBank mRNA | AK298441.1 | - | 5 | 1148 | ||
AK312968.1 | - | 6 | 1051 | |||
BC005046.1 | - | 6 | 1030 | 4040317 | ||
X03168.1 | - | 6 | 1011 |