Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077269.1 | NP_001070737.1 | 5 | 855 | ||
NM_003387.4 | NP_003378.3 | 5 | 791 | |||
XM_011511780.2 | XP_011510082.1 | 5 | 977 | |||
GenBank mRNA | AF031588.1 | - | 5 | 799 | ||
AF106062.1 | - | 1 | 120 | |||
AK301271.1 | - | 5 | 796 | |||
AK304194.1 | - | 5 | 808 | |||
BC110288.1 | - | 5 | 827 | 5184105 | ||
BQ441937.1 | - | 3 | 593 | |||
BX640870.1 | - | 5 | 855 | |||
X86019.1 | - | 5 | 895 |