Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003388.4 | NP_003379.3 | 3 | 463 | ||
NM_032421.2 | NP_115797.1 | 3 | 463 | |||
GenBank mRNA | AB006629.2 | - | 3 | 463 | ||
BC150332.1 | - | 3 | 474 | 8860506 |