Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030753.4 | NP_110380.1 | 3, 4 | 762 | ||
GenBank mRNA | AB067628.1 | - | 3, 4 | 613 | ||
AY009397.1 | - | 3, 4 | 718 | |||
BC112116.1 | - | 3, 4 | 620 | 8327584 | ||
BC112118.1 | - | 3, 4 | 620 | 8327586 | ||
BC114219.1 | - | 3, 4 | 617 | 5726751 |