Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058244.3 | NP_490645.1 | 1, 2 | 267 | ||
NR_125351.1 | 1, 2 | 267 | ||||
GenBank mRNA | AB057725.1 | - | 1, 2 | 74 | ||
AY009402.1 | - | 1, 2 | 169 |