Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003394.3 | NP_003385.2 | 4 | 1071 | ||
XM_011538722.1 | XP_011537024.1 | 3 | 469 | |||
XM_017019919.1 | XP_016875408.1 | 2 | 1103 | |||
GenBank mRNA | AB070724.1 | - | 3 | 675 | ||
AK312906.1 | - | 4 | 1075 | |||
BC096353.3 | - | 4 | 711 | 40010101 | ||
BC096354.1 | - | 4 | 711 | 40010102 | ||
BC096356.1 | - | 4 | 711 | 40010104 | ||
U81787.1 | - | 4 | 990 | |||
X97057.1 | - | 2 | 497 |