Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146218.1 | NP_001139690.1 | 5 | 452 | ||
NM_004627.4 | NP_004618.2 | 5 | 562 | |||
GenBank mRNA | AK293113.1 | - | 5 | 517 | ||
AK299144.1 | - | 5 | 405 | |||
AK310268.1 | - | 5 | 1066 | |||
BC012415.1 | - | 5 | 495 | 3865141 | ||
BC111393.1 | - | 5 | 451 | |||
DB498697.1 | - | 5 | 453 | |||
Y12478.1 | - | 5 | 506 |