Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079539.1 | NP_001073007.1 | 3 | 433 | ||
NM_005080.3 | NP_005071.2 | 3 | 433 | |||
GenBank mRNA | AB076383.1 | - | 3 | 393 | ||
AB076384.1 | - | 3 | 393 | |||
AK292188.1 | - | 3 | 430 | |||
AK313761.1 | - | 3 | 429 | |||
BC000938.1 | - | 3 | 390 | 3449394 | ||
BC012841.1 | - | 3 | 417 | 3856898 | ||
BC015709.1 | - | 3 | 326 | |||
BI545390.1 | - | 3 | 454 | |||
CR456611.1 | - | 3 | 388 | |||
M31627.1 | - | 3 | 394 |