Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141919.1 | NP_001135391.1 | 9, 10 | 674 | ||
NM_001141920.1 | NP_001135392.1 | 8, 9 | 632 | |||
NM_175569.2 | NP_780778.1 | 8, 9 | 629 | |||
XM_005274587.3 | XP_005274644.1 | 9, 10 | 1605 | |||
XM_017029787.1 | XP_016885276.1 | 8, 9 | 1577 | |||
GenBank mRNA | AF380356.1 | - | 8, 9 | 607 | ||
AF534880.1 | - | 8, 9 | 603 | |||
AK304164.1 | - | 9, 10 | 674 | |||
BC100764.2 | - | 9, 10 | 597 | |||
BC100765.2 | - | 9, 10 | 536 | 40000158 | ||
BC100766.2 | - | 8, 9 | 492 | 40000159 | ||
BC100767.1 | - | 8, 9 | 494 | 40000160 |