Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004628.4 | NP_004619.3 | 9 | 1383 | ||
NR_027299.1 | 8 | 1259 | ||||
XM_011534093.2 | XP_011532395.1 | 9 | 1340 | |||
GenBank mRNA | AK222844.1 | - | 8 | 1076 | ||
AK289761.1 | - | 9 | 1312 | |||
AK295711.1 | - | 9 | 1201 | |||
AK311039.1 | - | 8 | 1188 | |||
BC016620.1 | - | 9 | 1294 | 4509957 | ||
D21089.1 | - | 9 | 1384 | |||
EU530533.1 | - | 1 | 49 | |||
HM113488.1 | - | 9 | 1283 | |||
KJ534962.1 | - | 9 | 1279 | |||
KJ535085.1 | - | 8 | 1155 | |||
X65024.1 | - | 8 | 1119 |