Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004628.4 | NP_004619.3 | 13, 14 | 2530 | ||
NR_027299.1 | 12, 13 | 2406 | ||||
GenBank mRNA | AK222844.1 | - | 12, 13 | 2223 | ||
AK289761.1 | - | 13, 14 | 2459 | |||
AK295711.1 | - | 13, 14 | 2348 | |||
AK311039.1 | - | 12, 13 | 2336 | |||
BC016620.1 | - | 13, 14 | 2441 | 4509957 | ||
D21089.1 | - | 13, 14 | 2531 | |||
HM113488.1 | - | 13, 14 | 2430 | |||
KJ534962.1 | - | 13, 14 | 2426 | |||
KJ535085.1 | - | 11, 12 | 2142 | |||
X65024.1 | - | 12, 13 | 2266 |