Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003399.5 | NP_003390.4 | 4 | 518 | ||
GenBank mRNA | AK300954.1 | - | 4 | 539 | ||
BC126174.1 | - | 4 | 435 | 8991890 | ||
BC143901.1 | - | 4 | 435 | 9052419 | ||
BP270314.1 | - | 4 | 509 | |||
BX386512.2 | - | 4 | 518 | |||
CD617070.1 | - | 3 | 186 | |||
U90724.2 | - | 4 | 538 |