Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003407.3 | NP_003398.2 | 2 | 379 | ||
GenBank mRNA | AK314042.1 | - | 2 | 379 | ||
BC009693.1 | - | 2 | 361 | 3900312 | ||
M63625.1 | - | 2 | 330 | |||
M92843.1 | - | 2 | 380 |