Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021030.2 | NP_066358.2 | 4 | 1279 | ||
GenBank mRNA | AB021644.1 | - | 4 | 1229 | ||
AK074832.1 | - | 4 | 1264 | |||
AV726241.1 | - | 1 | 112 | |||
BC136323.1 | - | 4 | 1166 | 9020310 | ||
CD358782.1 | - | 1 | 399 |