Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006963.4 | NP_008894.2 | 2 | 302 | ||
GenBank mRNA | AK291508.1 | - | 2 | 204 | ||
AK312278.1 | - | 2 | 204 | |||
BC010642.1 | - | 2 | 192 | 3852749 | ||
BC026193.1 | - | 7 | 1268 | |||
BC041139.1 | - | 2 | 653 | 5751460 | ||
BC053687.1 | - | 2 | 196 | 5214077 | ||
BT009806.1 | - | 1 | 59 | |||
S50223.1 | - | 1 | 148 |