Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308123.1 | NP_001295052.1 | 3 | 635 | ||
NM_006965.3 | NP_008896.2 | 3 | 645 | |||
XM_005258341.3 | XP_005258398.1 | 3 | 632 | |||
GenBank mRNA | AF038964.1 | - | 3 | 589 | ||
AF542097.1 | - | 2 | 466 | |||
AK291246.1 | - | 3 | 624 | |||
BC003566.1 | - | 3 | 595 | 3537160 | ||
BC016801.1 | - | 3 | 616 | 4072599 | ||
BG436261.1 | - | 3 | 626 | |||
BT006658.1 | - | 2 | 454 | |||
CA314100.1 | - | 2 | 319 | |||
CR450328.1 | - | 2 | 454 | |||
U68536.1 | - | 3 | 618 |