Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005368.2 | NP_001005368.1 | 2 | 317 | ||
NM_001324164.1 | NP_001311093.1 | 2 | 179 | |||
NM_001324165.1 | NP_001311094.1 | 2 | 317 | |||
NM_001324166.1 | NP_001311095.1 | 2 | 179 | |||
NM_001324167.1 | NP_001311096.1 | 2 | 179 | |||
NM_001324168.1 | NP_001311097.1 | 2 | 317 | |||
NM_006973.2 | NP_008904.1 | 2 | 179 | |||
XM_017016616.1 | XP_016872105.1 | 1 | 3250 | |||
GenBank mRNA | AI871265.1 | - | 2 | 222 | ||
AK309483.1 | - | 2 | 186 | |||
AK312132.1 | - | 2 | 176 | |||
BC022842.1 | - | 1 | 114 | 5014207 | ||
BG477527.1 | - | 2 | 181 | |||
HY045180.1 | - | 2 | 318 | |||
HY148951.1 | - | 2 | 301 | |||
U69645.1 | - | 2 | 155 |
Gene Symbol: ZNF32-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047558.1 | 1 | 156 | |||
GenBank mRNA | BI459211.1 | - | 2 | 351 |