Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003420.3 | NP_003411.3 | 3 | 484 | ||
XM_017007129.1 | XP_016862618.1 | 3 | 458 | |||
XM_017007130.1 | XP_016862619.1 | 3 | 479 | |||
XM_017007131.1 | XP_016862620.1 | 3 | 480 | |||
GenBank mRNA | AK314821.1 | - | 3 | 483 | ||
BC013597.1 | - | 3 | 462 | 4186299 | ||
BP237260.1 | - | 3 | 484 | |||
BT006962.1 | - | 2 | 239 | |||
X07289.1 | - | 3 | 452 |